ADVH14DBCD2B AChemBlock

2'-Fluoro-biphenyl-3-amine

  • MDL number MFCD06802521
  • Cas Number : 176032-77-2
  • Molecular Weight : 187.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-fluoro-biphenyl-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40489
Availability details for
Quantity
  • Availability
  • Add to Cart