ADVH14DBCD52 AChemBlock

N-Boc-9-aminononan-1-ol

  • MDL number MFCD02094529
  • Cas Number : 1397043-36-5
  • Molecular Weight : 259.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-boc-9-aminononan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41392
Availability details for
Quantity
  • Availability
  • Add to Cart