ADVH14DBCD56 AChemBlock

N-(2-Aminoethyl)-2-methylpropanamide hydrochloride

  • MDL number MFCD17977074
  • Cas Number : 53673-16-8
  • Molecular Weight : 166.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-2-aminoethyl-2-methylpropanamide-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37576
Availability details for
Quantity
  • Availability
  • Add to Cart