ADVH14DBCD67 AChemBlock

2,4-Dichloro-5-methylquinazoline

  • MDL number MFCD11047029
  • Cas Number : 78052-20-7
  • Molecular Weight : 213.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-dichloro-5-methylquinazoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36085
Availability details for
Quantity
  • Availability
  • Add to Cart