ADVH14DBCD86 AChemBlock

(S)-3-(Cbz-amino)pyrrolidine hydrochloride

  • MDL number MFCD09749851
  • Cas Number : 1217631-74-7
  • Molecular Weight : 256.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-cbz-amino-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42226
Availability details for
Quantity
  • Availability
  • Add to Cart