ADVH14DBCD95 AChemBlock

tert-Butyl 2-(3-formylpiperidin-1-yl)-2-oxoethylcarbamate

  • MDL number MFCD28064151
  • Cas Number : 1803599-32-7
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-3-formylpiperidin-1-yl-2-oxoethylcarbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40392
Availability details for
Quantity
  • Availability
  • Add to Cart