ADVH14DBCDAE AChemBlock

Br-PEG4-OH

  • MDL number MFCD18205852
  • Cas Number : 85141-94-2
  • Molecular Weight : 257.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/br-peg4-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35794
Availability details for
Quantity
  • Availability
  • Add to Cart