ADVH14DBCDBA AChemBlock

BnO-PEG3-OH

  • MDL number MFCD02093556
  • Cas Number : 55489-58-2
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bno-peg3-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37447
Availability details for
Quantity
  • Availability
  • Add to Cart