ADVH14DBCDC6 AChemBlock

HO-PEG4-CH2COOtBu

  • MDL number MFCD27635170
  • Cas Number : 169751-72-8
  • Molecular Weight : 308.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ho-peg4-ch2cootbu.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40639
Availability details for
Quantity
  • Availability
  • Add to Cart