ADVH14DBCDD2 AChemBlock

FmocNH-PEG4-CH2COOH

  • MDL number MFCD26142981
  • Cas Number : 437655-95-3
  • Molecular Weight : 473.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/fmocnh-peg4-ch2cooh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38084
Availability details for
Quantity
  • Availability
  • Add to Cart