ADVH14DBCDD5 AChemBlock

H2N-PEG1-CH2CH2COOtBu

  • MDL number MFCD20727480
  • Cas Number : 1260092-46-3
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/h2n-peg1-ch2ch2cootbu.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42009
Availability details for
Quantity
  • Availability
  • Add to Cart