ADVH14DBCDDA AChemBlock

H2N-PEG6-CH2CH2COOtBu

  • MDL number MFCD13184944
  • Cas Number : 1286281-32-0
  • Molecular Weight : 409.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/h2n-peg6-ch2ch2cootbu.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41817
Availability details for
Quantity
  • Availability
  • Add to Cart