ADVH14DBCDE4 AChemBlock

3-[2-(Boc-amino)ethoxy]propanoic acid

  • MDL number MFCD22574796
  • Cas Number : 1260092-44-1
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-2-boc-amino-ethoxy-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42010
Availability details for
Quantity
  • Availability
  • Add to Cart