ADVH14DBCDED AChemBlock

BocNH-PEG7-CH2CH2COOH

  • MDL number MFCD29052162
  • Cas Number : 2055044-68-1
  • Molecular Weight : 497.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bocnh-peg7-ch2ch2cooh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39914
Availability details for
Quantity
  • Availability
  • Add to Cart