ADVH14DBCDEE AChemBlock

BocNH-PEG8-CH2CH2COOH

  • MDL number MFCD13185254
  • Cas Number : 1334169-93-5
  • Molecular Weight : 541.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bocnh-peg8-ch2ch2cooh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41646
Availability details for
Quantity
  • Availability
  • Add to Cart