ADVH14DBCE27 AChemBlock

2-Amino-5-hexynoic acid

  • MDL number MFCD09947719
  • Cas Number : 215160-72-8
  • Molecular Weight : 127.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-5-hexynoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39732
Availability details for
Quantity
  • Availability
  • Add to Cart