ADVH14DBCE38 AChemBlock

(S)-a-[Fmoc-(methyl)amino]cyclohexaneacetic acid

  • MDL number MFCD31704916
  • Cas Number : 925240-97-7
  • Molecular Weight : 393.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-a-fmoc-methyl-amino-cyclohexaneacetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35067
Availability details for
Quantity
  • Availability
  • Add to Cart