ADVH14DBCE3E AChemBlock

a-(Methylamino)cyclohexaneacetic acid

  • MDL number MFCD12047894
  • Cas Number : 1218286-14-6
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/a-methylamino-cyclohexaneacetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42206
Availability details for
Quantity
  • Availability
  • Add to Cart