ADVH14DBCE4C AChemBlock

(S)-2-(Fmoc-amino)-7-(Boc-amino)heptanoic acid

  • MDL number MFCD01860606
  • Cas Number : 194718-17-7
  • Molecular Weight : 482.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-fmoc-amino-7-boc-amino-heptanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40115
Availability details for
Quantity
  • Availability
  • Add to Cart