ADVH14DBCE68 AChemBlock

2-Methoxy-L-homophenylalanine

  • MDL number MFCD07372223
  • Cas Number : 1089276-09-4
  • Molecular Weight : 209.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methoxy-l-homophenylalanine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42710
Availability details for
Quantity
  • Availability
  • Add to Cart