ADVH14DBCE79 AChemBlock

2-Fluoro-O-methyl-L-tyrosine

  • MDL number MFCD18662191
  • Cas Number : 1270093-02-1
  • Molecular Weight : 213.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-fluoro-o-methyl-l-tyrosine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41870
Availability details for
Quantity
  • Availability
  • Add to Cart