ADVH14DBCE9D AChemBlock

N-Fmoc-N-methyl-D-aspartic acid 4-tert-butyl ester

  • MDL number MFCD04974458
  • Cas Number : 1799443-40-5
  • Molecular Weight : 425.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-fmoc-n-methyl-d-aspartic-acid-4-tert-butyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40405
Availability details for
Quantity
  • Availability
  • Add to Cart