ADVH14DBCEAF AChemBlock

tert-Butyl 5-iodopentylcarbamate

  • MDL number MFCD18384789
  • Cas Number : 262278-05-7
  • Molecular Weight : 313.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-iodopentylcarbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39253
Availability details for
Quantity
  • Availability
  • Add to Cart