ADVH14DBCEB9 AChemBlock

tert-Butyl (3-hydroxybutyl)carbamate

  • MDL number MFCD04974277
  • Cas Number : 167216-30-0
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-hydroxybutyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40708
Availability details for
Quantity
  • Availability
  • Add to Cart