ADVH14DBCEE3 AChemBlock

BocNH-PEG8-CH2CH2NH2

  • MDL number MFCD31656919
  • Cas Number : 1052207-59-6
  • Molecular Weight : 512.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bocnh-peg8-ch2ch2nh2.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42824
Availability details for
Quantity
  • Availability
  • Add to Cart