ADVH14DBCF0D AChemBlock

2-(4-Chlorophenyl)aziridine

  • MDL number MFCD21909712
  • Cas Number : 7763-73-7
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-chlorophenyl-aziridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36104
Availability details for
Quantity
  • Availability
  • Add to Cart