ADVH14DBCF32 AChemBlock

N-Boc-undecane-1,11-diamine

  • MDL number MFCD02094502
  • Cas Number : 937367-26-5
  • Molecular Weight : 286.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-boc-undecane-1-11-diamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P34995
Availability details for
Quantity
  • Availability
  • Add to Cart