ADVH14DBCF33 AChemBlock

N-Boc-nonane-1,9-diamine

  • MDL number MFCD02094500
  • Cas Number : 510754-90-2
  • Molecular Weight : 258.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-boc-nonane-1-9-diamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37752
Availability details for
Quantity
  • Availability
  • Add to Cart