ADVH14DBCF34 AChemBlock

1-Boc-decane-1,10-diamine

  • MDL number MFCD02094501
  • Cas Number : 216961-61-4
  • Molecular Weight : 272.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-boc-decane-1-10-diamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39694
Availability details for
Quantity
  • Availability
  • Add to Cart