ADVH14DBCF64 AChemBlock

Methyl 4-phenylpiperidine-4-carboxylate hydrochloride

  • MDL number MFCD11223623
  • Cas Number : 83898-15-1
  • Molecular Weight : 255.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-phenylpiperidine-4-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35876
Availability details for
Quantity
  • Availability
  • Add to Cart