ADVH14DBCF73 AChemBlock

(R)-tert-Butyl pyrrolidine-3-carboxylate

  • MDL number MFCD15475017
  • Cas Number : 681288-45-9
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-tert-butyl-pyrrolidine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36626
Availability details for
Quantity
  • Availability
  • Add to Cart