ADVH14DBCF7F AChemBlock

2-[2-(Hydroxymethyl)pyrrolidin-1-yl]acetonitrile

  • MDL number MFCD14660612
  • Cas Number : 1029698-92-7
  • Molecular Weight : 140.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-hydroxymethyl-pyrrolidin-1-yl-acetonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42944
Availability details for
Quantity
  • Availability
  • Add to Cart