ADVH14DBCF82 AChemBlock

mPEG6-OH

  • MDL number MFCD00045996
  • Cas Number : 23601-40-3
  • Molecular Weight : 296.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/mpeg6-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39483
Availability details for
Quantity
  • Availability
  • Add to Cart