ADVH14DBCF8E AChemBlock

HO-PEG6-CH2COOH

  • MDL number MFCD31656911
  • Cas Number : 120394-66-3
  • Molecular Weight : 340.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ho-peg6-ch2cooh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42315
Availability details for
Quantity
  • Availability
  • Add to Cart