ADVH14DBCF96 AChemBlock

BnO-PEG3-CH2COOH

  • MDL number MFCD31652691
  • Cas Number : 144751-75-7
  • Molecular Weight : 298.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bno-peg3-ch2cooh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41222
Availability details for
Quantity
  • Availability
  • Add to Cart