ADVH14DBCF9A AChemBlock

10-(Boc-amino)-1-decyl bromide

  • MDL number MFCD03788821
  • Cas Number : 887353-29-9
  • Molecular Weight : 336.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/10-boc-amino-1-decyl-bromide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35399
Availability details for
Quantity
  • Availability
  • Add to Cart