ADVH14DBCFA0 AChemBlock

mPEG5-CH2CH2COOH

  • MDL number MFCD20926383
  • Cas Number : 1347750-72-4
  • Molecular Weight : 324.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/mpeg5-ch2ch2cooh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41571
Availability details for
Quantity
  • Availability
  • Add to Cart