ADVH14DBCFAF AChemBlock

BocNH-PEG5-CH2CH2NH2

  • MDL number MFCD22056313
  • Cas Number : 189209-27-6
  • Molecular Weight : 380.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bocnh-peg5-ch2ch2nh2.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40218
Availability details for
Quantity
  • Availability
  • Add to Cart