ADVH14DBCFB0 AChemBlock

BocNH-PEG6-CH2CH2NH2

  • MDL number MFCD16619391
  • Cas Number : 1091627-77-8
  • Molecular Weight : 424.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bocnh-peg6-ch2ch2nh2.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42702
Availability details for
Quantity
  • Availability
  • Add to Cart