ADVH14DBCFB2 AChemBlock

11-(Boc-amino)-1-undecyl bromide

  • MDL number MFCD22576479
  • Cas Number : 463930-53-2
  • Molecular Weight : 350.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/11-boc-amino-1-undecyl-bromide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37979
Availability details for
Quantity
  • Availability
  • Add to Cart