ADVH14DBCFBD AChemBlock

BocNH-PEG11-OH

  • MDL number MFCD31656922
  • Cas Number : 1556847-53-0
  • Molecular Weight : 601.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bocnh-peg11-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40955
Availability details for
Quantity
  • Availability
  • Add to Cart