ADVH14DBCFC5 AChemBlock

BnO-PEG4-OH

  • MDL number MFCD06797175
  • Cas Number : 86259-87-2
  • Molecular Weight : 284.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bno-peg4-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35707
Availability details for
Quantity
  • Availability
  • Add to Cart