ADVH14DBCFC6 AChemBlock

BnO-PEG5-OH

  • MDL number MFCD06797154
  • Cas Number : 57671-28-0
  • Molecular Weight : 328.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bno-peg5-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37293
Availability details for
Quantity
  • Availability
  • Add to Cart