ADVH14DBCFC7 AChemBlock

BnO-PEG6-OH

  • MDL number MFCD06797107
  • Cas Number : 24342-68-5
  • Molecular Weight : 372.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bno-peg6-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39428
Availability details for
Quantity
  • Availability
  • Add to Cart