ADVH14DBCFD9 AChemBlock

mPEG8-OH

  • MDL number MFCD06797152
  • Cas Number : 25990-96-9
  • Molecular Weight : 384.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/mpeg8-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39285
Availability details for
Quantity
  • Availability
  • Add to Cart