ADVH14DBCFE0 AChemBlock

mPEG16-OH

  • MDL number MFCD27977525
  • Cas Number : 133604-58-7
  • Molecular Weight : 736.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/mpeg16-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41627
Availability details for
Quantity
  • Availability
  • Add to Cart