ADVH14DBD006 AChemBlock

H2N-PEG2-CH2COOH

  • MDL number MFCD13185897
  • Cas Number : 134978-97-5
  • Molecular Weight : 163.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/h2n-peg2-ch2cooh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41563
Availability details for
Quantity
  • Availability
  • Add to Cart