ADVH14DBD007 AChemBlock

H2N-PEG3-CH2COOH

  • MDL number MFCD24479841
  • Cas Number : 134978-99-7
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/h2n-peg3-ch2cooh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41562
Availability details for
Quantity
  • Availability
  • Add to Cart