ADVH14DBD021 AChemBlock

tert-Butyl 3-(2-bromoethoxy)propanoate

  • MDL number MFCD22574803
  • Cas Number : 1393330-36-3
  • Molecular Weight : 253.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-2-bromoethoxy-propanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41402
Availability details for
Quantity
  • Availability
  • Add to Cart