ADVH14DBD022 AChemBlock

Br-PEG2-CH2CH2COOtBu

  • MDL number MFCD22574822
  • Cas Number : 1381861-91-1
  • Molecular Weight : 297.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/br-peg2-ch2ch2cootbu.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41446
Availability details for
Quantity
  • Availability
  • Add to Cart